Anti-GFRA1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-BK5
10801-684EA 522.84 USD
10801-684
Anti-GFRA1 Rabbit Polyclonal Antibody
Antibodies
Glial cell line-derived neurotrophic factor (GDNF) is a potent survival factor for central and peripheral neurons and is essential for the development of kidneys and the enteric nerves system. Physiological responses to GDNF require the presence of a novel glycosylphosphadidylinositol linked protein GDNFRalpha, which is a cell surface receptor for GDNF. The cDNAs encoding GDNFRalpha from human, rat, chicken and mouse have been cloned recently. GDNFRalpha was also termed Ret ligand 1 (RETL1) or TGF-beta-related neurotrophic factor receptor 1 (TrnR1) and nominated as GFRa-1 recently. GFRa-1 binds GDNF specifically and mediates activation of the Ret protein tyrosine kinase (PTK). Thus, GDNF, GFRa and the Ret PTK form a complex to transduce GDNF signal and to mediate GDNF function.

Recommended Dilutions: ELISA: User Optimized; Immunohistochemsitry: 1 ?g/mL; Western Blot: User Optimized; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: GFRA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Internal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR