Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CFLAR Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

200-401-BF2
10802-896EA 423.98 USD InStock
10802-896
Anti-CFLAR Rabbit Polyclonal Antibody
Antibodies
Apoptosis is related to many diseases and induced by a family of cell death receptors and their ligands. Cell death signals are transduced by death domain (DD) containing adapter molecules and members of the ICE/CED-3 protease family.Caspases-8 (FLICE) and -10 (FLICE2) are two pivotal members in the ICE/CED-3 protease family. FLICE-inhibitory proteins were identified in virus and human and designated v-FLIPs and FLIP respectively. The human FLIP was also cloned by several labs independently and termed Casper, I-FLICE, FLAME-1, CASH, CLARP and MRIT. FLIP contains two death effector domains (DEDs) and a caspase-like domain. FLIP interacts with adapter protein FADD and caspase-8 and 10, and potently inhibits apoptosis induced by all known death receptors CD95, DR3, TRAIL-R and TNFR1.

Recommended Dilutions: ELISA: 1:5,000; Western Blot: 0.5 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CFLAR
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty