Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CCL11 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

200-401-BB0
10802-098EA 423.98 USD InStock
10802-098
Anti-CCL11 Rabbit Polyclonal Antibody
Antibodies
Chemokines play a key role in inflammation. The CC chemokine eotaxin is a potent and specific eosinophil chemoattractant that is expressed by a variety of cell types in certain inflammatory conditions. Some G-protein coupled chemokine receptors are also utilized as virus coreceptors for fusion and infection of cells. The eotaxin receptor CCR3 is required for HIV-1 entry into target cells such as microglia and eotaxin inhibits the infection of HIV-1.

Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: 10 μg/mL; Western Blot: 1 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CCL11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty