Anti-TNFRSF21 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-AX6
10802-546EA 518.29 USD
10802-546
Anti-TNFRSF21 Rabbit Polyclonal Antibody
Antibodies
Apoptosis is induced by certain cytokines including TNF and Fas ligand of the TNF family through their death domain containing receptors, TNF-R1 and Fas. Several novel death receptors including DR3, DR4, and DR5 were recently identified. A new death domain containing receptor in the TNFR family was cloned recently and termed DR6 for death receptor-6. Like TNF-R1, DR6 interacts with death domain containing adapter molecule TRADD. Overexpression of DR6 induces apoptosis and activates NF-kB and JNK. DR6 is widely expressed in human tissues and cell lines. The ligand for DR6 has not been identified.

Recommended Dilutions: ELISA: 1:5,000; Western Blot: 1:500; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: TNFRSF21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR