Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFRSF25 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

200-401-AX3
10800-994EA 423.98 USD InStock
10800-994
Anti-TNFRSF25 Rabbit Polyclonal Antibody
Antibodies
Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF and Fas ligand of the TNF family through their death domain containing receptors, TNFR1 and Fas. A novel cell death receptor was recently identified by several groups independently and designated DR3, Wsl-1, Apo-3, TRAMP and LARD1-5. The ligand for this novel death receptor has been defined as TWEAK, also termed Apo3L. DR3 is highly expressed in the tissues enriched in lymphocytes including PBL, thymus and spleen. Like TNFR1, DR3 induces apoptosis and NF-kB activation.

Recommended Dilutions: ELISA: 1:10,000; Immunofluorescence Microscopy: 20 μg/mL; Western Blot: 1 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: TNFRSF25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Extracellular
Host: Rabbit
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty