Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DFFA Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

200-401-AV5
10801-130EA 423.98 USD InStock
10801-130
Anti-DFFA Rabbit Polyclonal Antibody
Antibodies
Apoptosis is related to many diseases and induced by a family of cell death receptors and their ligands. Cell death signals are transduced by death domain containing adapter molecules and members of the caspase family of proteases. These death signals finally cause the degradation of chromosomal DNA by activated DNase. A human 45 kDa DNA fragmentation factor (DFF45) was identified recently that was cleaved by caspase-3 during apoptosis. Mouse homologue of human DFF45 was identified as a DNase inhibitor designated ICAD. DFF45/ICAD have short forms that were termed DFF35 and ICADs, respectively. Upon cleavage of DFF45/ICAD, the caspase activated deoxyribonuclease (DFF40/CAD) is released and activated and eventually causes the degradation of DNA in the nuclei. Therefore, the cleavage of DFF45/ICAD, which causes DFF40/CAD activation and DNA degradation, is the hallmark of apoptotic cell death.

Recommended Dilutions: ELISA: 1:10,000; Western Blot: 1:1,000; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: DFFA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty