Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ATF4 Mouse Monoclonal Antibody (ATTO 390) [clone: S360A-24]

Supplier: Rockland Immunochemical

200-367-W61 200-367-W61
10801-958EA 423.98 USD InStock
10801-958 CA10801-958
Anti-ATF4 Mouse Monoclonal Antibody (ATTO 390) [clone: S360A-24]
Antibodies
Cyclic AMP-dependent transcription factor ATF-4 (ATF4) is a basic leucine-zipper (bZip) transcription factor, which regulates amino acid metabolism, DNA damage repair, chromatin remodeling, and apoptosis in response to cellular and ER stress. ATF4 works with various proteins, such as C/EBP homology protein (CHOP), aspargine synthetase (ASNS), and cAMP response element (CRE) among others to mediate cellular stress. ATF4 also regulates glucose homeostasis by suppressing beta-cell proliferation and insulin production. Furthermore, ATF4 targets the histone demethylase JMJD3 to alter chromatin structure and enhance gene transcription in response to amino acid deprivation. Anti-ATF4 is ideal for researchers interested in Cell Signaling, Oncology, Cell Differentiation, and Apoptosis; relevant pathways include MAPK signaling pathways, Activation of cAMP-Dependent PKA, CREB pathways, GPCR pathways and Rho Family GTPases.

Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: User Optimized; Western Blot: 1:1000; 0.1% (w/v) Sodium Azide

Type: Primary
Antigen: ATF4
Clonality: Monoclonal
Clone: S360A-24
Conjugation: ATTO 390
Epitope: C-Terminal
Host: Mouse
Isotype:
Reactivity: Human, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty