Anti-FIG4 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: S202-7]

Supplier: Rockland Immunochemicals
200-365-W80
10801-412EA 464.71 USD
10801-412
Anti-FIG4 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: S202-7]
Fig4/Sac3 phosphatase, a 97-kDa protein, specifically removes the 5-phosphate from the inositol ring of PI(3,5)P2, a membrane-bound phospholipid, which acts as a molecular signal for trafficking and fusion of intracellular vesicles. It is broadly expressed in all eukaryotic cells and plays a vital role in intracellular membrane trafficking and signaling. Mutations in Fig4/Sac3 have been linked to pale tremor mice that exhibit neuropathy with severe central and peripheral neuronal degeneration, including the loss of neurons in the dorsal root ganglion and the sciatic nerve. Fig4/Sac3 knockout mice have been shown to be smaller in size and exhibit abnormal tremor and gait and neuronal autophagy. Anti-Fig4/Sac3 is ideal for research in Neuroscience and Cell Signaling.

Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: User Optimized; Western Blot: 1:1000; 0.1% (w/v) Sodium Azide

Type: Primary
Antigen: Sac3
Clonality: Monoclonal
Clone: S202-7
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope: C-terminal
Host: Mouse
Isotype:
Reactivity: Human, Rat, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR