Anti-ATXN1 Mouse Monoclonal Antibody (ATTO 594) [clone: S65-37]

Supplier: Rockland Immunochemicals
200-363-W87 200-363-W87
10800-134EA 433.54 USD
CA10800-134 10800-134
Anti-ATXN1 Mouse Monoclonal Antibody (ATTO 594) [clone: S65-37]
Antibodies
Ataxin-1 is a member of the ATXN1 protein family and contains a single AXH domain. It is a neurodegenerative disorder protein thought to have a role in the metabolism of RNA as it has been shown to localize to the RNA and transcription dependent inclusions within the nucleus. A mutation of Ataxin-1 is the cause of spinocerebellar ataxia type-1 (SCA1), a progressive, neurodegenerative disease that is autosomal dominant and primarily affects the Purjinke cells found in brain stem neuronal populations and the cerebellum. Expression of Ataxin-1 is almost ubiquitous, except in the brain where it is isolated to populations of neurons. Anti-Ataxin-1 is ideal for research in Neuroscience.

Recommended Dilutions: ELISA: 1:10,000; Western Blot: 1:1000; 0.1% (w/v) Sodium Azide

Type: Primary
Antigen: ATXN1
Clonality: Monoclonal
Clone: S65-37
Conjugation: ATTO 594
Epitope: Center
Host: Mouse
Isotype:
Reactivity: Human, Rat, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR