Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-6xHis Mouse Monoclonal Antibody (Cy5®) [clone: 33D1.D2.G8]

Supplier: Rockland Immunochemical

200-310-382 200-310-382
10799-510EA 434.58 USD InStock
10799-510 CA10799-510
Anti-6xHis Mouse Monoclonal Antibody (Cy5®) [clone: 33D1.D2.G8]
Antibodies
6X His Tag CY5 conjugated Antibody as well as other Epitope tags are short peptide sequences that are easily recognized by tag-specific antibodies.  Due to their small size, epitope tags do not affect the tagged protein’s biochemical properties.   Most often sequences encoding the epitope tag are included with target DNA at the time of cloning to produce fusion proteins containing the epitope tag sequence.  This allows anti-epitope tag antibodies to serve as universal detection reagents for any tag containing protein produced by recombinant means.  This means that anti-epitope tag antibodies are a useful alternative to generating specific antibodies to identify, immunoprecipitate or immunoaffinity purify a recombinant protein.  The anti-epitope tag antibody is usually functional in a variety of antibody-dependent experimental procedures.  Expression vectors producing epitope tag fusion proteins are available for a variety of host expression systems including bacteria, yeast, insect and mammalian cells.  Rockland Immunochemicals produces anti-epitope tag antibodies against many common epitope tags including Myc, GST, GFP, 6X His, MBP, FLAG and HA. Rockland Immunochemicals also produces antibodies to other tags including FITC, Rhodamine (TRITC), DNP and biotin.

Recommended Dilutions: FLISA: 1:10,000 - 1:50,000; Flow Cytometry: 1:500 - 1:2,500; Fluorochrome Protein Value: 10.4; Immunofluorescence Microscopy: 1:1,000 - 1:5,000 ; contains 0.01% (w/v) Sodium Azide

Type: Tag
Antigen: 6xHis
Clonality: Monoclonal
Clone: 33D1.D2.G8
Conjugation: Cy5®
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty