Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FIG4 Mouse Monoclonal Antibody (Biotin) [clone: S202-7]

Supplier: Rockland Immunochemical

200-306-W80
10802-894EA 423.98 USD InStock
10802-894
Anti-FIG4 Mouse Monoclonal Antibody (Biotin) [clone: S202-7]
Antibodies
Fig4/Sac3 phosphatase, a 97-kDa protein, specifically removes the 5-phosphate from the inositol ring of PI(3,5)P2, a membrane-bound phospholipid, which acts as a molecular signal for trafficking and fusion of intracellular vesicles. It is broadly expressed in all eukaryotic cells and plays a vital role in intracellular membrane trafficking and signaling. Mutations in Fig4/Sac3 have been linked to pale tremor mice that exhibit neuropathy with severe central and peripheral neuronal degeneration, including the loss of neurons in the dorsal root ganglion and the sciatic nerve. Fig4/Sac3 knockout mice have been shown to be smaller in size and exhibit abnormal tremor and gait and neuronal autophagy. Anti-Fig4/Sac3 is ideal for research in Neuroscience and Cell Signaling.

Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: User Optimized; Western Blot: 1:1000; 0.1% (w/v) Sodium Azide

Type: Primary
Antigen: Sac3
Clonality: Monoclonal
Clone: S202-7
Conjugation: Biotin
Epitope: C-terminal
Host: Mouse
Isotype:
Reactivity: Human, Rat, Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty