Anti-BCL2L11 Mouse Monoclonal Antibody [clone: 1C2H4]

Supplier: Rockland Immunochemicals
200-301-Z72
10800-374EA 528.48 USD
10800-374
Anti-BCL2L11 Mouse Monoclonal Antibody [clone: 1C2H4]
Antibodies
Members in the Bcl-2 family are critical regulators of apoptosis by either inhibiting or promoting cell death. Bcl-2 homology 3 (BH3) domain is a potent death domain. BH3 domain containing pro-apoptotic proteins, including Bad, Bax, Bid, Bik, and Hrk, form a growing subclass of the Bcl-2 family. Bim is another BH3 domain containing protein which can induce apoptosis. Bim interacts with diverse members in the pro-survival Bcl-2 sub-family including Bcl-2, Bcl-xL and Bcl-w. The messenger RNA of Bim is ubiquitously expressed in multiple tissues and cell lines.

Recommended Dilutions: ELISA: 1:10,000; Immunofluorescence Microscopy: 20 µg/mL; Western Blot: 1 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: BCL2L11
Clonality: Monoclonal
Clone: 1C2H4
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR