Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-APAF1 Mouse Monoclonal Antibody [clone: 5E11]

Supplier: Rockland Immunochemical

200-301-Y28 200-301-Y28
10800-048EA 423.98 USD InStock
10800-048 CA10800-048
Anti-APAF1 Mouse Monoclonal Antibody [clone: 5E11]
Antibodies
Apoptosis is related to many diseases and induced by a family of cell death receptors and their ligands. Cell death signals are transduced by death domain containing adapter molecules and members of the caspase family of proteases. The mammalian homologous of the key cell death gene CED-4 in C. elegans was identified recently from human and mouse and designated Apaf1 for apoptosis protease-activating factor 1. Apaf1 binds to cytochrome c (Apaf2) and caspase-9 (Apaf3), which leads to caspase-9 activation. Activated caspase-9 in turn cleaves and activates caspase-3 that is one of the key proteases, being responsible for the proteolytic cleavage of many key proteins in apoptosis. Apaf1 can also associate with caspase-4 and caspase-8. Apaf1 transcript is ubiquitously expressed in human tissues.

Recommended Dilutions: ELISA: User Optimized; Immunohistochemsitry: 0.5 μg/mL ; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: APAF1
Clonality: Monoclonal
Clone: 5E11
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty