Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RBFOX1 Mouse Monoclonal Antibody [clone: 1G10]

Supplier: Rockland Immunochemical

200-301-X08
10802-948EA 419.61 USD InStock
10802-948
Anti-RBFOX1 Mouse Monoclonal Antibody [clone: 1G10]
Antibodies
FOX1, also known as ataxin-2 binding protein 1 (A2BP1), is a neuron specific RNA-binding protein that has been shown to regulate a wide range of alternative splicing events implicated in neuronal development and maturation including transcription factors and synaptic proteins. Defects in FOX1 have also been associated with autism, attention-deficit hyperactivity disorder and schizophrenia. FOX1 Antibody is ideal for researchers interested in neuroscience research.

Recommended Dilutions: ELISA: 1:50,000; Immunofluorescence Microscopy: 1:500; Western Blot: 1:5000; 0.1% (w/v) Sodium Azide

Type: Primary
Antigen: RBFOX1
Clonality: Monoclonal
Clone: 1G10
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Mouse
Isotype:
Reactivity: Human, Bovine, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty