Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SPTAN1 Mouse Monoclonal Antibody [clone: 3D7]

Supplier: Rockland Immunochemical

200-301-W92 200-301-W92
10799-968EA 419.61 USD InStock
10799-968 CA10799-968
Anti-SPTAN1 Mouse Monoclonal Antibody [clone: 3D7]
Antibodies
The spectrin family of cytoskeletal proteins is comprised of 2 alpha genes (α1 and α2) and five beta genes (β1-β5). Spectrins have been shown to function as scaffolding proteins in mechanical support of the plasma membrane as well as bind other membrane proteins and lipids. Defects in spectrin genes have been linked to some forms of hereditary spherocytosis, a type of auto-hemolytic anemia which is characterized by spherical red blood cells that are more prone to lysis. Alpha II Spectrin Antibody is ideal for researchers interested in cell cycle research.

Recommended Dilutions: ELISA: 1:20,000; Immunofluorescence Microscopy: 1:500; Western Blot: 1:2000; 0.1% (w/v) Sodium Azide

Type: Primary
Antigen: SPTAN1
Clonality: Monoclonal
Clone: 3D7
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype:
Reactivity: Bovine, Human, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty