Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EZH2 Mouse Monoclonal Antibody

Supplier: Rockland Immunochemical

200-301-W46
10802-656EA 423.98 USD InStock
10802-656
Anti-EZH2 Mouse Monoclonal Antibody
Antibodies
EZH2 is a histone-lysine methyltransferase which methylates ‘Lys-9’ and ‘Lys-27’ of histone H3, leading to transcriptional repression. It is a member of the polycomb group (PcG) family which form multimeric protein complexes and are involved in maintaining the transcriptional repressive state of genes over successive cell generations. The EZH2 activity is dependent on the association with other components of the PRC2 complex (EED, SUZ12/JJAZ1, RBBP4 and RBBP7). EZH2 may play a role in the hematopoietic and internal nervous systems. Over-expression of EZH2 is observed during advanced stages of prostate cancer and breast cancer. Anti-EZH2 Antibody is ideal for research in Epigenetics, Gene Expression, Neuroscience and Cancer.

Recommended Dilutions: ChIP: 2.5 μg per IP ; contains 0.05% (w/v) Sodium Azide

Type: Primary
Antigen: EZH2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty