Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-5-mC Mouse Monoclonal Antibody [clone: 33D3]

Supplier: Rockland Immunochemical

200-301-V02 200-301-V02
10799-862EA 566.04 USD InStock
CA10799-862 10799-862
Anti-5-mC Mouse Monoclonal Antibody [clone: 33D3]
Antibodies
The 5-methylcytosine antibody (clone 33D3) is the most published and widely used antibody for DNA methylation analysis. Anti-5mC is ideal for research in Epigenetics due to its applicability to DNA methylation analysis.

Recommended Dilutions: ELISA: 1:3,000; ChIP: 1 μg/ChIP; Western Blot: 1:1,000; contains 0.05% (w/v) Sodium Azide

Type: Primary
Antigen: 5-mC
Clonality: Monoclonal
Clone: 33D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Vertebrates, Plants

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty