Anti-5-mC Mouse Monoclonal Antibody

Supplier: Rockland Immunochemicals
200-301-U98 200-301-U98
10799-864EA 528.48 USD
10799-864 CA10799-864
Anti-5-mC Mouse Monoclonal Antibody
Antibodies
5-methyl Cytidine is a modified nucleotide found in the DNA of both plants and vertebrates. Anti-5-mC Antibody is ideal for research in Epigenetics, specifically DNA methylation, due to its applicability to methylation analysis.

Recommended Dilutions: Immunofluorescence Microscopy: 1:500; Immunoprecipitation: 1-2 ?g/IP ; contains 0.05% (w/v) Sodium Azide

Type: Primary
Antigen: 5-mC
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Other (Wide Range)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR