Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GCG Mouse Monoclonal Antibody [clone: IMD-7]

Supplier: Rockland Immunochemical

200-301-P38
10801-744EA 423.98 USD InStock
10801-744
Anti-GCG Mouse Monoclonal Antibody [clone: IMD-7]
Antibodies
Glucagon is a member of a multigene family that includes secretin. Glucagon is a 29-amino acid pancreatic hormone that counteracts the glucose-lowering action of insulin by stimulating glycogenolysis and gluconeogenesis. the human glucagon gene is approximately 9.4 kb long and contains 6 exons and 5 introns, and assigned to 2q36-2q37. This antibody is suitable for researchers interested in cancer research, metabolic diseases, transcription factors and nuclear receptors, neuroscience research, SKT signaling, GCPR signaling, and calcium signaling research.

Recommended Dilutions: Immunohistochemsitry: 1:100-1:500 ; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: GCG
Clonality: Monoclonal
Clone: IMD-7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat, Rabbit

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty