Anti-GSN Mouse Monoclonal Antibody [clone: GEL-42]

Supplier: Rockland Immunochemicals
200-301-P37
10801-624EA 528.25 USD
10801-624
Anti-GSN Mouse Monoclonal Antibody [clone: GEL-42]
Antibodies
Gelsolin, a protein of leukocytes, platelets, and other cells, severs actin filaments in the presence of submicromolar calcium, thereby solating cytoplasmic actin gels. A gelsolin variant with 23 more N-terminal amino acids is a plasma component probably involved in the clearance of actin, the most abundant human protein, from the circulation. Gelsolin is located in 9q34. Plasma and cytoplasmic gelsolins are encoded by a single gene and contain a duplicated actin-binding domain. This antibody is suitable for researchers interested in cell research and cytoskeletal signaling research.

Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: 1:100-1:500; Western Blot: 0.5-1µg/ml; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: GSN
Clonality: Monoclonal
Clone: GEL-42
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rabbit Proteins.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR