Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DMD Mouse Monoclonal Antibody [clone: DYS-48]

Supplier: Rockland Immunochemical

200-301-P36
10801-212EA 423.98 USD InStock
10801-212
Anti-DMD Mouse Monoclonal Antibody [clone: DYS-48]
Antibodies
Dystrophin(DMD) gene has 79 exons spanning at least 2,300 kb (2.3 Mb). The C terminus of the dystrophin protein is encoded by a highly conserved, alternatively spliced region of the gene. beta-dystroglycan binding activity is expressed by the dystrophin fragment spanning amino acids 3026-3345 containing the ZZ domain. DMD transcript is formed by at least 60 exons; the first half of the transcript is formed by a minimum of 33 exons spanning nearly 1000 kb, and the remaining portion has at least 27 exons that may spread over a similar distance. Dystrophin gene is expressed at a higher level in primary cultures of neuronal cells than in astro-glial cells derived from adult mouse brain. overexpression of dystrophin prevents the development of the abnormal mechanical properties associated with dystrophic muscle without causing deleterious side effects. This antibody is suitable for researchers interested in Muscular Dystrophies and Dystrophin-Glycoprotein Complexes and cardiovascular research.

Recommended Dilutions: ELISA: 1:20,000; Immunohistochemsitry: 1:100-1:500; Western Blot: 1-2ug/ml; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: DMD
Clonality: Monoclonal
Clone: DYS-48
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat, Rabbit

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty