Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DES Mouse Monoclonal Antibody [clone: DES-82]

Supplier: Rockland Immunochemical

200-301-P35
10802-440EA 423.98 USD InStock
10802-440
Anti-DES Mouse Monoclonal Antibody [clone: DES-82]
Antibodies
Desmin belongs to the type III family of intermediate filaments, a class of cytoskeletal elements. DES gene encodes desmin, a muscle-specific cytoskeletal protein found in smooth, cardiac, and heart muscles. Tidball (1992) found that desmin was codistributed with actin thin filaments within the cellular processes of myotendinous junctions in frog skeletal muscle. DES gene contains 9 exons and spans about 8.4 kb. By in situ hybridization, Viegas-Pequignot et al. (1989) localized the gene to 2q35. Desmin mutation responsible for idiopathic dilated cardiomyopathy. This antibody is suitable for researchers interested in cancer research, cardiovascular diseases, neurodegenerative diseases, cell adhesion research, and cytoskeletal signaling research.

Recommended Dilutions: ELISA: 1:20,000; Immunohistochemsitry: 1:100-1:500; Western Blot: 2ug/ml; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: DES
Clonality: Monoclonal
Clone: DES-82
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat, Rabbit

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty