Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CK7 Mouse Monoclonal Antibody [clone: CK-7]

Supplier: Rockland Immunochemical

200-301-P34
10801-078EA 423.98 USD InStock
10801-078
Anti-CK7 Mouse Monoclonal Antibody [clone: CK-7]
Antibodies
KRT7 is a type II keratin of simple nonkeratinizing epithelia. The deduced 489-amino acid protein has a calculated molecular mass of about 54 kD. K7 contains 4 central alpha-helical segments with heptad repeats of hydrophobic residues characteristic of a coiled-coil region. Within this domain, K7 shares 73% homology with epidermal K6B. KRT7 gene contains 9 exons and spans more than 15.6 kb. K7 gene is mapped to chromosome 12. Keratin 7 is expressed in a wide range of epithelial structures in humans. This antibody is suitable for researchers interested in cancer research, cardiovascular diseases, and cytoskeletal signaling research.

Recommended Dilutions: ELISA: 1:20,000; Immunohistochemsitry: 1:100-1:500; Western Blot: 1-2ug/ml; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: CK7
Clonality: Monoclonal
Clone: CK-7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty