Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CK18 Mouse Monoclonal Antibody [clone: CK-18]

Supplier: Rockland Immunochemical

200-301-P33
10801-076EA 423.98 USD InStock
10801-076
Anti-CK18 Mouse Monoclonal Antibody [clone: CK-18]
Antibodies
Intermediate filaments (IFs) are a structurally related family of cellular proteins that appear to be intimately involved with the cytoskeleton. Human keratin 18(KRT18) and the homologous mouse Endo B are type I IF protein subunits whose expression is restricted in adults to a variety of simple epithelial tissues. The KRT18 gene is 3,791 bp long and the keratin 18 protein is coded for by 7 exons. The K18 gene is 3791 bp in length and the K18 protein is coded for by seven exons. By Southern blotting using the genomic DNA PCR product, the gene for keratin 18 is assigned to chromosome 12. Mutation of human keratin 18 in association with cryptogenic cirrhosis. This antibody is suitable for researchers interested in cancer research, cardiovascular diseases, and cytoskeletal signaling research.

Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: 1:100-1:500; Western Blot: 0.5-1µg/ml; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: CK18
Clonality: Monoclonal
Clone: CK-18
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty