Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CDC6 Mouse Monoclonal Antibody [clone: IMD-6]

Supplier: Rockland Immunochemical

200-301-P29
10800-718EA 423.98 USD InStock
10800-718
Anti-CDC6 Mouse Monoclonal Antibody [clone: IMD-6]
Antibodies
In yeasts, Cdc6 (Saccharomyces cerevisiae) and Cdc18 (Schizosaccharomyces pombe) associate with the origin recognition complex (ORC) proteins to render cells competent for DNA replication. Cdc6 is overexpressed in human cancers, where it has a critical regulatory role in addition to DNA replication. Yan et al. (1998) showed that Cdc6 is expressed selectively in proliferating but not quiescent mammalian cells, both in culture and within tissues in intact animals. During the transition from a growth-arrested to a proliferative state, transcription of mammalian Cdc6 is regulated by E2F proteins, as revealed by a functional analysis of the human Cdc6 promoter and by the ability of exogenously expressed E2F proteins to stimulate the endogenous Cdc6 gene. They conclude that expression of human Cdc6 is regulated in response to mitogenic signals though transcriptional control mechanisms involving E2F proteins, and that Cdc6 is required for initiation of DNA replication in mammalian cells. This antibody is suitable for researchers interested in cancer research and Cell Cycle research.

Recommended Dilutions: ELISA: 1:20,000; Western Blot: 1-2µg/ml; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: CDC6
Clonality: Monoclonal
Clone: IMD-6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty