Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CALM1 Mouse Monoclonal Antibody [clone: CM-16]

Supplier: Rockland Immunochemical

200-301-P25
10800-504EA 423.98 USD InStock
10800-504
Anti-CALM1 Mouse Monoclonal Antibody [clone: CM-16]
Antibodies
Calmodulin is the archetype of the family of calcium-modulated proteins of which nearly 20 members have been found. Calmodulin contains 149 amino acids and has 4 calcium-binding domains. Its functions include roles in growth and the cell cycle as well as in signal transduction and the synthesis and release of neurotransmitters. Three calmodulin genes (CALM1, CALM2, and CALM3) map to chromosomes 14q24-q31, 2p21.1-p21.3, and 19q13.2-q13.3, respectively. This antibody is suitable for researchers interested in cardiovascular diseases, neurodegenerative diseases, autoimmune diseases, Cancer research, Developmental Signaling, Inflammatory Signaling, Cytoskeletal Signaling, GPCR signaling, AKT Signaling, and MAK Kinase Signaling.

Recommended Dilutions: ELISA: 1:5000; Western Blot: 0.5µg/ml; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: CALM1
Clonality: Monoclonal
Clone: CM-16
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty