Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PPP3R1 Mouse Monoclonal Antibody [clone: CC-6]

Supplier: Rockland Immunochemical

200-301-P23
10800-498EA 434.58 USD InStock
10800-498
Anti-PPP3R1 Mouse Monoclonal Antibody [clone: CC-6]
Antibodies
Calcineurin, the Ca(2+)/calmodulin-regulated protein phosphatase, first detected in skeletal muscle and brain, has been found in all cells from yeast to mammals. Calcineurin A alpha (PPP3CA),is located on human chromosomes 4, Chromosomal mapping of the human genes for the calmodulin-dependent protein phosphatase (calcineurin) catalytic subunit. Calcineurin regulates bone formation by the osteoblast. This antibody is suitable for researchers interested in apoptosis, transcription factors, nuclear receptors, cancer research, Immune Response, cardiovascular disease, MAK Kinase Signaling, and AKT Signaling.

Recommended Dilutions: ELISA: 1:5000; Immunohistochemsitry: 1:100-1:500; Western Blot: 0.25-0.5ug/ml; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: PPP3R1
Clonality: Monoclonal
Clone: CC-6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat, Bovine

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty