The human calbindin D28k (CALB1) is a calcium-binding protein belonging to the troponin C superfamily. It was originally described as a 27-kD protein induced by vitamin D in the duodenum of the chick. Parmentier et al. (1991) mapped the CALB1 gene to 8q21.3-q22.1 by in situ hybridization. This antibody is suitable for researchers interested in transcription factors, nuclear receptors, cancer research, and nutritional and metobolic diseases.
Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: 1:100-1:500; Western Blot: 0.5-1ug/ml; contains 0.01% (w/v) Sodium Azide
Type: Primary
Antigen: CALB1
Clonality: Monoclonal
Clone: CB-D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human