Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BAP1 Mouse Monoclonal Antibody [clone: BP-1]

Supplier: Rockland Immunochemical

200-301-P21
10801-998EA 423.98 USD InStock
10801-998
Anti-BAP1 Mouse Monoclonal Antibody [clone: BP-1]
Antibodies
BAP1, also known as BRCA1-associated protein-1, contains an acidic region, a highly charged C-terminal region, and 2 putative nuclear localization signals. BAP1 is a novel ubiquitin hydrolase which binds to the BRCA1 RING finger and enhances BRCA1-mediated cell growth suppression. BAP1 is expressed as a 4-kb mRNA in all human tissues, and mapped to 3p21.3. This antibody is suitable for researchers interested in Epigenetics, transcription factors, nuclear receptors, cancer research, and ubiquitin research.

Recommended Dilutions: ELISA: 1:5000; Immunohistochemsitry: 1:100-1:500; Western Blot: 0.5µg/ml; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: BAP1
Clonality: Monoclonal
Clone: BP-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty