Anti-CD4 Mouse Monoclonal Antibody [clone: 9H5A8]

Supplier: Rockland Immunochemicals
200-301-AJ0
10800-678EA 528.48 USD
10800-678
Anti-CD4 Mouse Monoclonal Antibody [clone: 9H5A8]
Antibodies
CD4 is a member of the immunoglobulin superfamily and is implicated as associative recognition element in MHC (Major Histocompatibility Complex) class II-restricted immune response. On T-lymphocytes, it defines the helper/inducer subset. The mature 55 kd CD4 protein consists of a 372 amino acid extracellular segment composed of four tandem immunoglobulin-like VJ regions. The CD4 molecule is a major receptor for human immunodeficiency virus (HIV), binding directly to the envelope glycoprotein gp120 on HIV, with the co-receptors being CCR5 or CXCR4. It has been shown that the V-like domains are critical for binding with HIV envelope gp120.

Recommended Dilutions: ELISA: 1:500-1:10,000; Western Blot: 0.5-1 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 9H5A8
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR