Anti-Biotin Mouse Monoclonal Antibody [clone: 4C7.D5]

Supplier: Rockland Immunochemicals
200-301-098L
10802-066EA 2616.59 USD
10802-066
Anti-Biotin Mouse Monoclonal Antibody [clone: 4C7.D5]
Antibodies
Biotin Antibody detects Biotin. Biotin is a water-soluble B-complex vitamin (vitamin B7). It is composed of a ureido (tetrahydroimidizalone) ring fused with a tetrahydrothiophene ring. A valeric acid substituent is attached to one of the carbon atoms of the tetrahydrothiophene ring. Biotin is a coenzyme for carboxylase enzymes, involved in the synthesis of fatty acids, isoleucine, and valine, and in gluconeogenesis. Biotin is necessary for cell growth, the production of fatty acids, and the metabolism of fats and amino acids. Anti-Biotin Antibody is ideal for investigators involved in Cell Signaling and Cell Biology research.

Recommended Dilutions: ELISA: 1:20,000; Flow Cytometry: 1-2 µg/mL; Immunohistochemsitry: 1:1,000 - 1:5,000; Western Blot: 1:2,000 - 1:10,000; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: Biotin
Clonality: Monoclonal
Clone: 4C7.D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR