Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ACHE Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical

200-106-232S 200-106-232S
10799-538EA 124.57 USD InStock
CA10799-538 10799-538
Anti-ACHE Goat Polyclonal Antibody (Biotin)
Antibodies
Acetyl Cholinesterase is abundantly expressed in erythrocytes and is part of the type-B carboxylesterase/lipase family. Anti-Acetyl Cholinesterase cancels signal transduction at the neuromuscular junction via hydrolysis of acetylcholine as it is released into the synaptic cleft by the motor neurons. Anti-Acetyl Cholinesterase antibody is ideal for investigators involved in Neuroscience and Neurotransmitter research.

Recommended Dilutions: ELISA: 1:2,000 - 1:10,000; Immunoprecipitation: 1:100; Western Blot: 1:500 - 1:2,500; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: ACHE
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty