Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GDHB Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical

200-106-214S
10801-748EA 124.57 USD InStock
10801-748
Anti-GDHB Goat Polyclonal Antibody (Biotin)
Antibodies
Anti-Glucose Dehydrogenase antibody recognizes glucose dehydrogenase. A member of the oxidoreductase family that acts on CH-OH group on donors, glucose dehydrogenase uses NAD+ or NADP+ as the acceptor. Glucose dehyrogenase reacts with beta-D-glucose to produce D-glucono-1,5-lactone and depending on the acceptor, either NADH or NADPH. Anti-Glucose Dehydrogenase is ideal for investigators interested in Metabolism, Cancer, and Signal Transduction.

Recommended Dilutions: ELISA: 1:5,000 - 1:25,000; Western Blot: 1:500 - 1:3,000; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: gdhB
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Bacillus

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty