Anti-choA Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
200-106-213S
10802-378EA 147.5 USD
10802-378
Anti-choA Goat Polyclonal Antibody (Biotin)
Antibodies
Anti-Cholesterol Oxidase antibody is a member of the GMC oxidoreductase family. Cholesterol Oxidase acts as a catalyst in the oxidation of the 3-beta-hydroxy group of cholesterol and the isomerization of the double bond of new product. Anti-Cholesterol Oxidase is ideal for investigators interested in Metabolism, Signal Transduction, or Cardiovascular research.

Recommended Dilutions: ELISA: 1:5,000 - 1:20,000; Immunoprecipitation: 1:100; Western Blot: 1:500 - 1:5,000; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: choA
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR