Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Bilirubin Oxidase Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical

200-106-212S
10800-370EA 124.57 USD InStock
10800-370
Anti-Bilirubin Oxidase Goat Polyclonal Antibody (Biotin)
Antibodies
Anti-Bilirubin Oxidase antibody is an enzyme that increases the oxidation rate of bilirubin to biliverdin and other substrates. It is ideal for investigators interested in Metabolism or Signal Transduction research.

Recommended Dilutions: ELISA: 1:1,000 - 1:5,000; Immunoprecipitation: 1:100; Western Blot: 1:500 - 1:2,500; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: Bilirubin Oxidase
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Myrothecium Verrucaria

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty