Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GLPK Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical

200-106-211S
10801-752EA 124.57 USD InStock
10801-752
Anti-GLPK Goat Polyclonal Antibody (Biotin)
Antibodies
Anti-Glycerol Kinase antibody recognizes the phosphotransferase enzyme glycerol kinase. Glycerol kinase catalyzes the formation of glycerol phosphate by transferring a phosphate from ATP to glycerol. Dihydroxyactone and L-glycerladehyde can also act as acceptors in place of glycerol. Anti-Glycerol Kinase is ideal for investigators interested in Cancer, Metabolism, and Kinase/Phosphatase enzymology.

Recommended Dilutions: ELISA: 1:1,000 - 1:5,000; Western Blot: 1:500 - 1:2,500; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: glpK
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Glycerol Kinase [Cellulomonas Species]

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty