Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GPD1 Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical

200-106-210S
10801-756EA 124.57 USD InStock
10801-756
Anti-GPD1 Goat Polyclonal Antibody (Biotin)
Antibodies
Anti-Glycerol-3-Phosphate Dehydrogenase recognizes the enzyme glycerol-3-phosphate dehydrogenase, a major component of lipid biosynthesis. Glyercol-3-phosphate dehydrogenase catalyzes the reduction of dihydroxyacetone phosphate (DHAP) to glycerol-3-phosphate. In addition, it assists in maintaining the redox potential across the inner mitochondrial membrane in glycloysis. Glycerol-3-phosphate dehydrogenase can be found in the cytosol and the inner mitochondrial membrane. Anti-Glycerol-3-Phosphate Dehydrogenase is ideal for investigators interested in metabolism, cancer, and cardiovascular diseases.

Recommended Dilutions: ELISA: 1:1,000 - 1:5,000; Western Blot: 1:500 - 1:2,500; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: GPD1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty