Anti-A2M Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
200-106-207S 200-106-207S
10799-978EA 147.5 USD
10799-978 CA10799-978
Anti-A2M Goat Polyclonal Antibody (Biotin)
Antibodies
Anti-alpha-2-Macroglobulin antibody is secreted into the plasma and belongs to the protease inhibitor I39 family. Anti-alpha-2-Macroglobulin inhibits all proteinases utilizing a “bait region,” or specific cleavage site, which traps the proteinase, after which a thioester bond is hydrolyzed and regulates the covalent binding of the protein to proteinase. Anti-alpha-2-Macroglobulin antibody is ideal for investigators interested in Cardiovascular , Protease Inhibitor, or Cell Biology research.

Recommended Dilutions: ELISA: 1:20,000 - 1:100,000; Western Blot: 1:2,000 - 1:10,000; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: A2M
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR