Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FOLR1 Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical

200-106-148S
10801-446EA 124.57 USD InStock
10801-446
Anti-FOLR1 Goat Polyclonal Antibody (Biotin)
Antibodies
Anti-Folate Binding Protein antibody belongs to the folate receptor family and binds to folate and reduced folic acid derivatives. It further regulates delivery of 5-methyltetrahydrofolate to the cell interior. Folate expresses solely in epithelial tissues, and expression is increased in malignant tissues. Anti-Folate Binding Protein antibody is ideal for researchers investigating Tags & Cell Markers and Cancer research.

Recommended Dilutions: ELISA: 1:1,000 - 1:2,500; Immunoprecipitation: 1:100; Western Blot: 1:500 - 1:1,000; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: FOLR1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty