Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SERPINA1 Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical

200-106-147S 200-106-147S
10799-974EA 124.57 USD InStock
10799-974 CA10799-974
Anti-SERPINA1 Goat Polyclonal Antibody (Biotin)
Antibodies
A member of the serpin family, Anti-Alpha-1-Anti-Trypsin inhibits serine proteases. With a moderate affinity for plasmin and thrombin, Anti-Alpha-1-Anti-Trypsin antibody mainly targets elastase. It inhibits trypsin, chymotrypsin and plasminogen activator by inhibiting insulin-induced NO synthesis. It also decreases coagulation time and has proteolytic activity against insulin and plasmin. The short peptide from AAT also targets elastase, but not trypsin. Anti-Alpha-1-Anti-Trypsin is ideal for researchers involved in Metabolism, Cancer, and Cardiovascular research.

Recommended Dilutions: ELISA: 1:50,000; Fluorochrome Protein Value: 10-20; Western Blot: 1:2,000 - 1:10,000; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: SERPINA1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty