Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Choline Oxidase Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical

200-106-094S
10802-382EA 124.57 USD InStock
10802-382
Anti-Choline Oxidase Goat Polyclonal Antibody (Biotin)
Antibodies
Choline Oxidase is a flavoprotein responsible for the catalyzing the conversion of choline to betaine aldehyde in animals and bacteria. Anti-Choline Oxidase antibody is ideal for researchers interested in Metabolism and Signal Transduction research.

Recommended Dilutions: ELISA: 1:1,000 - 1:2,500; Immunoprecipitation: 1:100; Western Blot: 1:500 - 1:1,000; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: Choline Oxidase
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Alcaligenes Species

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty