Anti-ABC3735 Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
200-106-092S 200-106-092S
10800-038EA 147.5 USD
10800-038 CA10800-038
Anti-ABC3735 Goat Polyclonal Antibody (Biotin)
Antibodies
Uricase is a peroxisomal enzyme that catalyzes the conversion of urea to allantoin. Uricase performs this action by cleaving the purine ring of uric acid rendering it much more soluble within the body for excretion. Uricase within the Bacillus species is of high importance due to its high activity and thermostability over a wide range of pH's. Anti-Uricase Antibody is ideal for investigators in Cell Biology and Microbiology research.

Recommended Dilutions: ELISA: 1:1,000 - 1:2,500; Immunoprecipitation: 1:100; Western Blot: 1:500 - 1:1,000; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: ABC3735
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Bacillus Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR