Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SPTAN1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

100-401-W92 100-401-W92
10799-966EA 419.61 USD InStock
CA10799-966 10799-966
Anti-SPTAN1 Rabbit Polyclonal Antibody
Antibodies
The spectrin family of cytoskeletal proteins is comprised of 2 alpha genes (α1 and α2) and five beta genes (β1-β5). Spectrins have been shown to function as scaffolding proteins in mechanical support of the plasma membrane as well as bind other membrane proteins and lipids (Bennett and Baines 2001). Defects in spectrin genes have been linked to some forms of hereditary spherocytosis, a type of auto-hemolytic anemia which is characterized by spherical red blood cells that are more prone to lysis. Alpha II Spectrin Antibody is ideal for researchers interested in cell cycle research.

Recommended Dilutions: ELISA: 1:50,000; Immunofluorescence Microscopy: 1:500; Western Blot: 1:5000;

Type: Primary
Antigen: SPTAN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Rat, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty