Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BRD2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

100-401-V88
10802-260EA 262.26 USD InStock
10802-260
Anti-BRD2 Rabbit Polyclonal Antibody
Antibodies
Brd2, a mitogen-activated kinase localized to the nucleus, is a putative transcriptional regulator which interacts with E2F1 and with histone H4 acetylated at ‘Lys-13’. It is expressed during development and may be involved in growth control. Brd2 also may play a role in spermatogenesis or folliculogenesis and may be involved in some types of leukemia and in juvenile myoclonic epilepsy. Anti-Brd2 Antibody is ideal for research Gene Expression, Cell Cycle Control and Cancer.

  • Recommended Dilutions: ELISA: 1:100; Western Blot: 1:1,000; contains 0.05% (w/v) Sodium Azide

Type: Primary
Antigen: BRD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty