Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ASH2L Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

100-401-V87 100-401-V87
10800-096EA 423.98 USD InStock
CA10800-096 10800-096
Anti-ASH2L Rabbit Polyclonal Antibody
Antibodies
Ash2 is a component of the Set1/Ash2 histone methyltransferase (HMT) complex. This complex specifically methylates K4 of histone H3, thereby activating transcription. Methylation of K4 is blocked by premethylation of the neighboring K9, a repressor of transcription. This indicates that the Set1/Ash2 HMT complex mediates the crosstalk between K9 methylation and K4 methylation. Ash2 plays a role in hematopoiesis and may be associated with some kinds of leukemia. Anti-Ash2 Antibody is ideal for research in Gene Expression, Epigenetics, Cell Signaling and Cancer.

Recommended Dilutions: ELISA: 1:100 - 1:500; Immunofluorescence Microscopy: 1:200; Western Blot: 1:1,000; contains 0.05% (w/v) Sodium Azide

Type: Primary
Antigen: ASH2L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty