Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RTF1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

100-401-V38
10801-196EA 423.98 USD InStock
10801-196
Anti-RTF1 Rabbit Polyclonal Antibody
Antibodies
PAF is a five-subunit protein complex composed of Paf1, Cdc73, Leo1, Rtf1 and Ctr9, which was first purified from yeast. The yeast PAF (yPAF) complex interacts with RNA polymerase II and coordinates the setting of histone marks associated with active transcription. The human PAF (hPAF) complex shares four subunits with yPAF (hCtr9, hPaf1, hLeo1 and Cdc73), but contains an additional higher eukaryotic-specific subunit, hSki8. In addition to coordinating events during transcription (initiation, promoter clearance and elongation); PAF also coordinates events in RNA quality control. Anti-dRtf1 Antibody is ideal for research in Gene Expression, Transcription and Epigenetics.

Recommended Dilutions: ChIP: 1-5 μg/ChIP; Immunohistochemsitry: 1:50; Immunoprecipitation: 12 μg per IP; Western Blot: 1:1,000; contains 0.05% (w/v) Sodium Azide

Type: Primary
Antigen: RTF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Drosophila, Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty