Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RUNX1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

100-401-V33 100-401-V33
10799-994EA 423.98 USD InStock
10799-994 CA10799-994
Anti-RUNX1 Rabbit Polyclonal Antibody
Antibodies
This antibody specifically recognizes the AML1 (RUNX1) (UniProtKB/Swiss-Prot entry Q01196) - ETO (RUNX1T1) (UniProtKB/Swiss-Prot entry Q06455) fusion protein that arises due to a translocation between chromosome 8 and 22. This translocation is one of the most frequent karyotypic abnormalities observed in acute myeloid leukaemia. It produces a chimerical gene made up of the 5’-region of AML1and the 3’-region of ETO. The chimerical protein is thought to associate with the nuclear corepressor/histone deacetylase complex to block hematopoietic differentiation. Anti-AML1-ETO Antibody is ideal for research in Cancer and Gene Expression.

Recommended Dilutions: ELISA: 1:500; ChIP: 4 μl/ChIP; Western Blot: 1:1,000; contains 0.05% (w/v) Sodium Azide and 0.05% ProClin 300

Type: Primary
Antigen: RUNX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty