Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CMKLR1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

100-401-V32
10800-616EA 423.98 USD InStock
10800-616
Anti-CMKLR1 Rabbit Polyclonal Antibody
Antibodies
CBFb represents the beta subunit of a heterodimeric core-binding transcription factor belonging to the PEBP2/CBF transcription factor family. These transcription factors regulate a host of genes specific to haematopoiesis (e.g. RUNX1) and osteogenesis (e.g. RUNX2). The beta subunit is the regulatory subunit which allosterically enhances the activity of the DNA binding alpha subunit as the complex binds to the core site of various enhancers and promoters. CBFb can be involved in a chromosomal rearrangement of chromosome 16 (inv(16)(p13q22)) which produces a fusion protein consisting of the N terminus of CBFb and the C-terminal portion of MYH11. This chromosomal rearrangement is associated with acute myeloid leukaemia of the M4Eo subtype. Anti-CBFb Antibody is ideal for research in Gene Expression, Cell Differentiation and Cancer.

Recommended Dilutions: ELISA: 1:500; ChIP: 4 μl/ChIP ; contains 0.05% (w/v) Sodium Azide

Type: Primary
Antigen: CMKLR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty