Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RUNX1T1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

100-401-V08
10802-636EA 423.98 USD InStock
10802-636
Anti-RUNX1T1 Rabbit Polyclonal Antibody
Antibodies
ETO is a transcriptional regulator which belongs to the myeloid translocation gene family. ETO exerts its function by interaction with transcription factors bound to promoters and binding to histone deacetylases. It recruits a range of corepressors to facilitate transcriptional repression. The t(8;21)(q22;q22) translocation is one of the most frequent karyotypic abnormalities in acute myeloid leukaemia. This translocation produces a chimeric gene made up of the 5’-region of AML1 and the 3’-region of the ETO gene. The chimeric protein is thought to associate with the nuclear corepressor/histone deacetylase complex to block hematopoietic differentiation. Anti-ETO Antibody is ideal for research in Gene Expression, Transcription and Cancer.

Recommended Dilutions: ELISA: 1:100; ChIP: 4 μl/ChIP ; contains 0.05% (w/v) Sodium Azide

Type: Primary
Antigen: RUNX1T1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty