Would you like to log in again?
VWR® Pipet Tips, Standard Line
Supplier: VWR International
10017-216 10017-038 10017-226 10017-036 10017-214 10017-218 10017-196 10017-204 10017-194 10017-202 10017-086 10017-208 10017-220 10017-034 10017-212 10017-198 10017-210 10017-200 10017-224 10017-222 10017-084 10017-206
10017-216CS 326.17 USD InStock
10017-216 10017-038 10017-226 10017-036 10017-214 10017-218 10017-196 10017-204 10017-194 10017-202 10017-086 10017-208 10017-220 10017-034 10017-212 10017-198 10017-210 10017-200 10017-224 10017-222 10017-084 10017-206
VWR® Pipet Tips, Standard Line
Pipet Tips
VWR® Pipet Tips, Standard Line are conveniently racked in the most widely used style of rack on the market or available in reload and bulk packaging.
  • Translucent packaging for easy vizualization of tips
  • Built with stability for ease of use when multi-channel pipetting
  • Back hinge makes it easy to open and close with one hand
  • Certified free of DNA, DNase, RNase, and endotoxins (pyrogens)
  • Manufactured under stringent controls in an ISO 9001 certified facility
  • Every lot is tested for precision, assuring maximum quality and compatibility
  • All raw materials are traceable
The 200µL tips are also available in the Rack & Stack style.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty